8-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A358111-10G A358111-1G A358111-250MG A358111-5G
77195-286EA 816.79 USD
77195-286 77195-288 77195-290 77195-292
8-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine 97%
8-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.47 g/mol
MDL Number: MFCD22683352
CAS Number: 1433822-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR