(R)-2-Methylpyrrolidine hydrochloride 98%

Supplier: Ambeed
Warning

A358074-1G A358074-250MG A358074-25G A358074-5G
77167-782EA 16 USD
77167-782 77167-784 77167-786 77551-012
(R)-2-Methylpyrrolidine hydrochloride 98%
(R)-2-Methylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN
MW: 121.6085 g/mol
MDL Number: MFCD06796610
CAS Number: 135324-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR