Fmoc-3-chloro-L-tyrosine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Tyr(3-Cl)-OH

A357911-1G A357911-250MG A357911-5G
77195-280EA 20 USD
77195-280 77195-282 77195-284
Fmoc-3-chloro-L-tyrosine 95%
Fmoc-3-chloro-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClNO₅
MDL Number: MFCD02682261
CAS Number: 478183-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR