1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 97%

Supplier: Ambeed
Warning

A355834-100MG A355834-1G A355834-250MG A355834-5G
77167-750EA 20.6 USD
77167-750 77167-752 77167-754 77167-756
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid 97%
1,4-Dioxaspiro[4.5]decane-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MW: 186.205 g/mol
MDL Number: MFCD11847571
CAS Number: 66500-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR