1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%

Supplier: Ambeed
Warning

A354411-10G A354411-25G A354411-100G A354411-1000G
77195-244EA 6 USD
77195-244 77616-550 77746-696 77746-695
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

Precaución: for Research Use Only

Formula: C₈H₁₆Cl₆O₃
MDL Number: MFCD02179352
CAS Number: 6001-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR