1-(2-Methoxyphenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-(O-Methoxyphenyl)ethanol

A354014-1G A354014-250MG A354014-5G
77195-238EA 20 USD
77195-238 77195-240 77195-242
1-(2-Methoxyphenyl)ethanol 98%
1-(2-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00004512
CAS Number: 13513-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR