Pentan-3-ylhydrazine hydrochloride 95%

Supplier: Ambeed
Danger

A352766-100MG A352766-1G A352766-250MG
77167-736EA 36.05 USD
77167-736 77167-738 77167-740
Pentan-3-ylhydrazine hydrochloride 95%
Pentan-3-ylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₅ClN₂
MW: 138.63 g/mol
MDL Number: MFCD14583114
CAS Number: 221024-42-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR