3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine, 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 3-(Pyrrolidinomethyl)phenylboronic acid, pinacol ester

A352526-1G A352526-250MG A352526-5G
77195-218EA 94 USD
77195-218 77195-220 77195-222
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 95%
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₂
MW: 287.205 g/mol
MDL Number: MFCD08741425
CAS Number: 884507-45-3
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR