1,4-Difluoro-2,3-dimethylbenzene 98%

Supplier: Ambeed
Danger

Synonyms: 3,6-Difluoro-o-xylene

A352505-250MG A352505-25G A352505-5G
77195-212EA 38.11 USD
77195-212 77195-214 77195-216
1,4-Difluoro-2,3-dimethylbenzene 98%
1,4-Difluoro-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂
MW: 142.146 g/mol
MDL Number: MFCD04973769
CAS Number: 1736-90-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR