1-(5-Fluoropyridin-2-yl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A352182-1G A352182-250MG
77195-208EA 419.21 USD
77195-208 77195-210
1-(5-Fluoropyridin-2-yl)cyclopropanecarboxylic acid 97%
1-(5-Fluoropyridin-2-yl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈FNO₂
MW: 181.16 g/mol
MDL Number: MFCD20646242
CAS Number: 1302580-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR