1-Benzyl-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A350340-100MG A350340-1G A350340-250MG
77167-732EA 56 USD
77167-732 77892-720 77892-721
1-Benzyl-1H-pyrrole-2-carboxylic acid 97%
1-Benzyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD00894448
CAS Number: 18159-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR