1-(2-Hydroxy-6-(trifluoromethyl)pyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

A350288-100MG A350288-1G A350288-250MG
77167-726EA 19 USD
77167-726 77167-728 77167-730
1-(2-Hydroxy-6-(trifluoromethyl)pyridin-3-yl)ethanone 95%
1-(2-Hydroxy-6-(trifluoromethyl)pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.13 g/mol
MDL Number: MFCD09754105
CAS Number: 116548-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR