N-Succinimidyl tetradecanoate 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl tetradecanoate, N-Succinimidyl Myristate, Tetradecanoic acid N-Succinimidyl ester, NHS-Myristate, Myristic acid N-Succinimidyl ester

A349514-1G A349514-250MG A349514-25G
77167-714EA 24.01 USD
77167-714 77167-716 77892-719
N-Succinimidyl tetradecanoate 95%
N-Succinimidyl tetradecanoate

Precaución: for Research Use Only

Formula: C₁₈H₃₁NO₄
MDL Number: MFCD00522592
CAS Number: 69888-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR