tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A349477-1G A349477-250MG A349477-25G A349477-5G
77167-706EA 43 USD
77167-706 77167-708 77167-710 77167-712
tert-Butyl (1-(4-bromophenyl)ethyl)carbamate 95%
tert-Butyl (1-(4-bromophenyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MDL Number: MFCD11506012
CAS Number: 850363-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR