1,4-Diethynylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: p-Diethynylbenzene, 1,4-Diethylnylbenzene

A349474-1G A349474-250MG A349474-25G A349474-5G
77195-200EA 22 USD
77195-200 77195-202 77195-204 77195-206
1,4-Diethynylbenzene 98%
1,4-Diethynylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₆
MW: 126.16 g/mol
MDL Number: MFCD00078375
CAS Number: 935-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR