(R)-tert-Butyl 3-(quinazolin-4-ylamino)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A349196-100MG A349196-250MG
77195-192EA 101.97 USD
77195-192 77195-194
(R)-tert-Butyl 3-(quinazolin-4-ylamino)pyrrolidine-1-carboxylate 97%
(R)-tert-Butyl 3-(quinazolin-4-ylamino)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₄O₂
MW: 314.38 g/mol
MDL Number: MFCD21602209
CAS Number: 1365937-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR