6-Chloro-4-methylpyridazin-3-amine 97%

Supplier: Ambeed
Warning

A349037-100MG A349037-1G A349037-250MG A349037-25G A349037-5G
77167-696EA 15.45 USD
77167-696 77167-698 77167-700 77167-702 77167-704
6-Chloro-4-methylpyridazin-3-amine 97%
6-Chloro-4-methylpyridazin-3-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.574 g/mol
MDL Number: MFCD19203440
CAS Number: 64068-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR