4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine 95%
Supplier: Ambeed
|
Warning
|
A348563-100MG
A348563-1G
A348563-250MG
77167-690EA
80
USD
77167-690
77167-692
77167-694
4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine 95%
4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine
Precaución: for Research Use Only
Formula:
C₇H₆ClF₃N₂O |
MDL Number:
MFCD23703477 CAS Number: 156425-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...