Uridine-5′-monophosphate disodium hydrate 97%

Supplier: Ambeed
Warning

Synonyms: 5'-Uridylic acid disodium salt xhydrate, Uridylic acid disodium salt hydrate, 5'-Uridylic acid disodium salt hydrate, U 5′-P hydrate, UMP hydrate, disodium [(2R,3S,4S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate hydrate, Uridine 5‘-monophosphate sel disodique hydrate, 5′-UMP-Na2 hydrate

A348462-100G A348462-25G A348462-500G
77167-682EA 18 USD
77167-682 77167-684 77167-686 77167-688
Uridine-5′-monophosphate disodium hydrate 97%
Uridine-5′-monophosphate disodium hydrate

Precaución: for Research Use Only

Formula: C₉H₁₁N₂Na₂O₉P·xH₂O
MW: 386.160 g/mol
MDL Number: MFCD00149422
CAS Number: 681435-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR