Potassium (3-bromophenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: potassium (3-bromophenyl)trifluoroborate, (3-Bromophenyl)trifluoroboric acid potassium salt

A347606-100G A347606-25G A347606-500G A347606-5G
77167-664EA 180 USD
77167-664 77167-666 77167-668 77167-670
Potassium (3-bromophenyl)trifluoroborate 98%
Potassium (3-bromophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₄BBrF₃K
MW: 262.91 g/mol
MDL Number: MFCD02093977
CAS Number: 374564-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR