7-Bromo-1H-indole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-3-formyl-1H-indole, 7-Bromo-1H-indole-3-carbaldehyde

A347555-1G A347555-250MG A347555-5G
77167-658EA 54.59 USD
77167-658 77167-660 77167-662
7-Bromo-1H-indole-3-carboxaldehyde 98%
7-Bromo-1H-indole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.054 g/mol
MDL Number: MFCD06657151
CAS Number: 115666-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR