2-Methoxypropan-1-ol 98%

Supplier: Ambeed
Danger

A347508-10G A347508-25G A347508-5G
77167-652EA 10 USD
77167-652 77167-654 77167-656
2-Methoxypropan-1-ol 98%
2-Methoxypropan-1-ol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MDL Number: MFCD00126948
CAS Number: 1589-47-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR