5-(Dimethylamino)pyridazin-3(2H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A347412-100MG A347412-250MG
77167-648EA 16.48 USD
77167-648 77167-650
5-(Dimethylamino)pyridazin-3(2H)-one hydrochloride 97%
5-(Dimethylamino)pyridazin-3(2H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃O
MW: 175.62 g/mol
MDL Number: MFCD28252257
CAS Number: 41773-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR