3-Ethylbenzothiazolium bromide 98+%

Supplier: Ambeed
Warning

A347205-100G A347205-1G A347205-25G A347205-500G A347205-5G
77167-640EA 231 USD
77167-640 77167-642 77167-644 77167-646 77612-980
3-Ethylbenzothiazolium bromide 98+%
3-Ethylbenzothiazolium bromide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNS
MDL Number: MFCD00051852
CAS Number: 32446-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR