2-Oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylic acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxo-1,2-dihydro-1,8-naphthyridine-3-carboxylate

A346880-1G A346880-250MG
77167-636EA 60.77 USD
77167-636 77167-638
2-Oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylic acid ethyl ester 95%
2-Oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD06659016
CAS Number: 5174-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR