(4-(Piperidin-1-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A345787-250MG A345787-1G
77195-174EA 26 USD
77195-174 77616-516
(4-(Piperidin-1-yl)phenyl)boronic acid 97%
(4-(Piperidin-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₂
MDL Number: MFCD03095171
CAS Number: 229009-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR