6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A345727-100MG A345727-1G A345727-250MG
77167-618EA 64.89 USD
77167-618 77167-620 77167-622
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinolin-2(1H)-one 97%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₃
MDL Number: MFCD18427677
CAS Number: 1207370-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR