Methyl 5-cyano-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A345274-10G A345274-1G A345274-250MG A345274-25G A345274-5G
77167-608EA 78 USD
77167-608 77167-610 77167-612 77167-614 77167-616
Methyl 5-cyano-2-methylbenzoate 95%
Methyl 5-cyano-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD18399111
CAS Number: 103261-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR