(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((benzoyloxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl benzoate 97%

Supplier: Ambeed
Warning

A344812-10G A344812-1G A344812-250MG A344812-5G A344812-100G
77167-586EA 28.84 USD
77167-586 77167-588 77167-590 77167-592 77684-021
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((benzoyloxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl benzoate 97%
(2R,3R,4R,5R)-5-(4-Benzamido-2-oxopyrimidin-1(2H)-yl)-2-((benzoyloxy)methyl)-4-fluoro-4-methyltetrahydrofuran-3-yl benzoate

Precaución: For research use only.

Formula: C₃₁H₂₆FN₃O₇
MW: 571.55 g/mol
MDL Number: MFCD23106344
CAS Number: 817204-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR