Ethyl 4-(((methyl(phenyl)amino)methylene)amino)benzoate 97%

Supplier: Ambeed
Warning

A344403-100G A344403-10G A344403-25G A344403-500G
77167-572EA 64.89 USD
77167-572 77167-574 77167-576 77167-578
Ethyl 4-(((methyl(phenyl)amino)methylene)amino)benzoate 97%
Ethyl 4-(((methyl(phenyl)amino)methylene)amino)benzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂
MW: 282.33 g/mol
MDL Number: MFCD09263624
CAS Number: 57834-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR