Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate 95%

Supplier: Ambeed
Warning

A343757-100G A343757-10G A343757-1G
77167-562EA 164.8 USD
77167-562 77167-564 77167-566
Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate 95%
Methyl 3-((2,2-dimethylbutanoyl)thio)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₃S
MW: 218.31 g/mol
MDL Number: MFCD22200648
CAS Number: 938063-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR