6-Nitrobenzo[d]isoxazole 98%

Supplier: Ambeed
Warning

A343127-100MG A343127-250MG
77167-556EA 27.81 USD
77167-556 77167-558
6-Nitrobenzo[d]isoxazole 98%
6-Nitrobenzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
MDL Number: MFCD06659638
CAS Number: 39835-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR