Promethazine hydrochloride 99%

Supplier: Ambeed
Danger

Synonyms: 10-[2-(Dimethylamino)propyl]phenothiazine Hydrochloride, N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine hydrochloride

A342357-1MG A342357-250MG
77195-160EA 5.15 USD
77195-160 77195-162
Promethazine hydrochloride 99%
Promethazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₂S
MW: 320.88 g/mol
MDL Number: MFCD00012652
CAS Number: 58-33-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR