2-(4-(4-Cyanostyryl)styryl)benzonitrile 97%

Supplier: Ambeed
Warning

A342092-100G A342092-5G
77195-156EA 66 USD
77195-156 77195-158 77746-670
2-(4-(4-Cyanostyryl)styryl)benzonitrile 97%
2-(4-(4-Cyanostyryl)styryl)benzonitrile

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₂
MDL Number: MFCD08458271
CAS Number: 13001-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR