(S)-Methyl 2-amino-3-(tritylthio)propanoate 95%

Supplier: Ambeed
Warning

A341489-1G A341489-250MG
77195-154EA 142 USD
77195-154 77746-667
(S)-Methyl 2-amino-3-(tritylthio)propanoate 95%
(S)-Methyl 2-amino-3-(tritylthio)propanoate

Precaución: for Research Use Only

Formula: C₂₃H₂₃NO₂S
MDL Number: MFCD27578273
CAS Number: 115545-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR