5-Formyl-2-hydroxybenzonitrile 97%

Supplier: Ambeed
Warning

A341332-100MG A341332-1G A341332-250MG
77195-148EA 38 USD
77195-148 77195-150 77195-152
5-Formyl-2-hydroxybenzonitrile 97%
5-Formyl-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MDL Number: MFCD16999751
CAS Number: 73289-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR