Potassium trifluoro(4-methylbenzyl)borate 95%

Supplier: Ambeed
Warning

A341162-100MG A341162-1G A341162-250MG
77195-142EA 29 USD
77195-142 77195-144 77195-146
Potassium trifluoro(4-methylbenzyl)borate 95%
Potassium trifluoro(4-methylbenzyl)borate

Precaución: for Research Use Only

Formula: C₈H₉BF₃K
MDL Number: MFCD09993057
CAS Number: 1422539-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR