6-Chloro-4-methyl-1,2,3,4-tetrahydroisoquinoline 97%
Supplier: Ambeed
|
Warning
|
A340710-100MG
A340710-10G
A340710-1G
A340710-250MG
A340710-5G
77195-132EA
42.23
USD
77195-132
77195-134
77195-136
77195-138
77195-140
6-Chloro-4-methyl-1,2,3,4-tetrahydroisoquinoline 97%
6-Chloro-4-methyl-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClN MW: 181.66 g/mol |
MDL Number:
MFCD12755840 CAS Number: 73037-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...