[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic Acid 95%

Supplier: Ambeed
Warning

A339989-1G A339989-5G
77167-530EA 10.3 USD
77167-530 77167-532 77167-534 77167-536
[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic Acid 95%
[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic Acid

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₄
MDL Number: MFCD00389774
CAS Number: 13653-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR