Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98+%

Supplier: Ambeed
Warning

A339958-1G A339958-250MG A339958-5G
77167-524EA 89.61 USD
77167-524 77167-526 77167-528
Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98+%
Methyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄
MW: 277.124 g/mol
MDL Number: MFCD22494137
CAS Number: 844500-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR