Levothyroxine sodium 95%

Supplier: Ambeed
Warning

Synonyms: 3,3′,5,5′-Tetraiodo-L-thyronine monosodium salt, Sodium O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine, Levoxyl, Sodium (S)-2-amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoate xhydrate, L-Thyroxine sodium salt

A338272-1G A338272-5G
77167-496EA 8 USD
77167-496 77167-498 77167-500
Levothyroxine sodium 95%
Levothyroxine sodium

Precaución: for Research Use Only

Formula: C₁₅H₁₀I₄NNaO₄·xH₂O
MW: 798.85 g/mol (anhy)
MDL Number: MFCD06408007
CAS Number: 25416-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR