Ethyl imidazo[1,2-a]pyrazine-3-carboxylate 97%

Supplier: Ambeed
Warning

A338132-100MG A338132-250MG
77195-122EA 24.72 USD
77195-122 77195-124
Ethyl imidazo[1,2-a]pyrazine-3-carboxylate 97%
Ethyl imidazo[1,2-a]pyrazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
MDL Number: MFCD28142909
CAS Number: 1286754-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR