2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dihydroxyphenyl)-2-(4-hydroxyphenyl)ethanone

A337993-100MG A337993-250MG A337993-1G
77195-118EA 16.48 USD
77195-118 77195-120 77746-657
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone 95%
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.246 g/mol
MDL Number: MFCD00498162
CAS Number: 17720-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR