2,6-Dimethoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

A337920-1G A337920-250MG
77195-114EA 33.99 USD
77195-114 77195-116
2,6-Dimethoxy-3-nitropyridine 97%
2,6-Dimethoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MW: 184.149 g/mol
MDL Number: MFCD00832858
CAS Number: 18677-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR