Methyl (4-nitro-1H-pyrazol-1-yl)acetate 98%

Supplier: Ambeed
Warning

A337672-250MG A337672-5G
77195-112EA 22 USD
77195-112 77746-656
Methyl (4-nitro-1H-pyrazol-1-yl)acetate 98%
Methyl (4-nitro-1H-pyrazol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₄
MDL Number: MFCD00297173
CAS Number: 6715-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR