1-Methyl-3-phenyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A336711-100MG A336711-250MG
77195-094EA 35 USD
77195-094 77746-655
1-Methyl-3-phenyl-1H-pyrazole 97%
1-Methyl-3-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MDL Number: MFCD00462160
CAS Number: 3463-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR