3-Amino-N-methylbenzamide 95%

Supplier: Ambeed
Warning

A335836-100G A335836-25G A335836-5G A335836-1G
77195-068EA 381 USD
77195-068 77195-070 77195-072 77616-480
3-Amino-N-methylbenzamide 95%
3-Amino-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD00070630
CAS Number: 25900-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR