1-(4-Nitrophenylazo)-2-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: Para Red, 1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol

A335314-25G A335314-5G A335314-1G
77195-064EA 44 USD
77195-064 77195-066 77746-654
1-(4-Nitrophenylazo)-2-naphthol 97%
1-(4-Nitrophenylazo)-2-naphthol

Precaución: for Research Use Only

Formula: C₁₆H₁₁N₃O₃
MDL Number: MFCD00003908
CAS Number: 6410-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR