(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-3-D-Pal-OH, 3-(Pyridin-3-yl)-D-alanine, (2R)-2-Amino-3-(pyridin-3-yl)propionic acid, 3-Aza-D-phenylalanine

A334849-1G A334849-250MG A334849-5G
77167-472EA 53.56 USD
77167-472 77167-474 77167-476
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid 98%
(2R)-2-Amino-3-(pyridin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00079673
CAS Number: 70702-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR