2,3,5,6-Tetrafluorophenyl trifluoroacetate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,3,5,6-Tetrafluorophenyl 2,2,2-trifluoroacetate, 2,3,5,6-Tetrafluorophenyl 2,2,2-trifluoroethanoate, Trifluoroacetic acid 2,3,5,6-tetrafluorophenyl ester
A334488-1G
A334488-250MG
A334488-25G
A334488-5G
77167-458EA
55.62
USD
77167-458
77167-460
77167-462
77683-999
2,3,5,6-Tetrafluorophenyl trifluoroacetate 95%
2,3,5,6-Tetrafluorophenyl trifluoroacetate
Precaución: For research use only.
Formula:
C₈HF₇O₂ MW: 262.081 g/mol |
MDL Number:
MFCD07784266 CAS Number: 142685-25-4 UN: 3272 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...